(2-Fluoro-6-methoxyphenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A378267-1G A378267-250MG A378267-5G
77168-040EA 39.14 USD
77168-040 77168-042 77168-044
(2-Fluoro-6-methoxyphenyl)methanamine hydrochloride 97%
(2-Fluoro-6-methoxyphenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀FNO·ClH
MW: 191.63 g/mol
MDL Number: MFCD12546099
CAS Number: 1214366-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR