4-Chloro-5-fluoroindoline-2,3-dione 97%

Supplier: Ambeed
Warning

A378157-250MG A378157-1G
77195-578EA 23.69 USD
77195-578 77746-756
4-Chloro-5-fluoroindoline-2,3-dione 97%
4-Chloro-5-fluoroindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₃ClFNO₂
MW: 199.57 g/mol
MDL Number: MFCD03618550
CAS Number: 84378-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR