Methyl 3-((4-(((tert-butoxycarbonyl)amino)methyl)phenyl)sulfonyl)propanoate 95%

Supplier: Ambeed
Warning

A377967-100MG A377967-1G A377967-250MG
77168-034EA 40.17 USD
77168-034 77168-036 77168-038
Methyl 3-((4-(((tert-butoxycarbonyl)amino)methyl)phenyl)sulfonyl)propanoate 95%
Methyl 3-((4-(((tert-butoxycarbonyl)amino)methyl)phenyl)sulfonyl)propanoate

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₆S
MW: 357.42 g/mol
MDL Number: MFCD27957005
CAS Number: 1362160-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR