2-Bromo-6-methyl-4-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A376840-100MG A376840-1G A376840-250MG
77195-550EA 124.63 USD
77195-550 77195-552 77195-554
2-Bromo-6-methyl-4-nitrobenzoic acid 97%
2-Bromo-6-methyl-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MW: 260.04 g/mol
MDL Number: MFCD28745203
CAS Number: 1807209-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR