Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide 95+%

Supplier: Ambeed
Warning

A376028-100MG A376028-1G A376028-250MG
77168-026EA 33.99 USD
77168-026 77168-028 77168-030
Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide 95+%
Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide

Precaución: for Research Use Only

Formula: C₁₀H₁₃BNO₃·Li
MW: 212.967 g/mol
MDL Number: MFCD16038156
CAS Number: 1014717-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR