2,6-Dimethylnicotinic acid 98%

Supplier: Ambeed
Warning

A375211-25G A375211-5G A375211-1G
77168-022EA 299 USD
77168-022 77168-024 77892-774
2,6-Dimethylnicotinic acid 98%
2,6-Dimethylnicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD06738677
CAS Number: 5860-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR