3-(1,1-Difluoroethyl)aniline 95%

Supplier: Ambeed
Danger

A375124-1G A375124-250MG
77168-018EA 187.46 USD
77168-018 77168-020
3-(1,1-Difluoroethyl)aniline 95%
3-(1,1-Difluoroethyl)aniline

Precaución: for Research Use Only

Formula: C₈H₉F₂N
MDL Number: MFCD20483887
CAS Number: 40788-05-4
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR