1-(5-Nitropyridin-2-yl)piperazine 95%

Supplier: Ambeed
Warning

A375108-1G A375108-250MG A375108-5G
77195-540EA 19 USD
77195-540 77195-542 77195-544
1-(5-Nitropyridin-2-yl)piperazine 95%
1-(5-Nitropyridin-2-yl)piperazine

Precaución: for Research Use Only

Formula: C₉H₁₂N₄O₂
MDL Number: MFCD00053059
CAS Number: 82205-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR