1-Methyl-4-(methylamino)piperidine 95%

Supplier: Ambeed
Danger

A375069-10G A375069-1G A375069-25G A375069-5G
77168-010EA 104.03 USD
77168-010 77168-012 77168-014 77168-016
1-Methyl-4-(methylamino)piperidine 95%
1-Methyl-4-(methylamino)piperidine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MDL Number: MFCD00006498
CAS Number: 73579-08-5
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR