2-Hydrazinopyrazine 95%

Supplier: Ambeed
Warning

A374904-100G A374904-10G A374904-1G A374904-25G
77167-996EA 331.66 USD
77167-996 77167-998 77168-000 77168-002
2-Hydrazinopyrazine 95%
2-Hydrazinopyrazine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MW: 110.12 g/mol
MDL Number: MFCD04114555
CAS Number: 54608-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR