3-Acetyl-2-chloropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-acetyl-2-chloropyridine, 1-(2-Chloro-3-pyridinyl)-1-ethanone, 1-(2-Chloropyridin-3-yl)ethanone

A374835-100G A374835-10G A374835-1G A374835-25G A374835-5G
77167-986EA 319.3 USD
77167-986 77167-988 77167-990 77167-992 77167-994
3-Acetyl-2-chloropyridine 97%
3-Acetyl-2-chloropyridine

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.5816 g/mol
MDL Number: MFCD03840751
CAS Number: 55676-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR