(2-(Methoxymethoxy)-5-((trifluoromethyl)thio)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A373989-100MG A373989-1G A373989-250MG
77195-514EA 35.02 USD
77195-514 77195-516 77195-518
(2-(Methoxymethoxy)-5-((trifluoromethyl)thio)phenyl)boronic acid 97%
(2-(Methoxymethoxy)-5-((trifluoromethyl)thio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BF₃O₄S
MW: 282.04 g/mol
MDL Number: MFCD18837607
CAS Number: 2096331-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR