(S)-(+)-2-Chloro-1-phenylethanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Chloro-1-phenylethanol, (S)-(+)-alpha-(Chloromethyl)benzyl alcohol

A373723-100G A373723-25G A373723-5G
77195-508EA 170 USD
77195-508 77195-510 77195-512
(S)-(+)-2-Chloro-1-phenylethanol 97%
(S)-(+)-2-Chloro-1-phenylethanol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MDL Number: MFCD00077692
CAS Number: 70111-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR