5-Iodo-2-(4-morpholino)pyridine 98%
Supplier: Ambeed
|
Warning
|
A372266-250MG
A372266-100MG
A372266-1G
A372266-5G
77167-972EA
20
USD
77167-972
77892-769
77892-770
77892-771
5-Iodo-2-(4-morpholino)pyridine 98%
5-Iodo-2-(4-morpholino)pyridine
Precaución: for Research Use Only
Formula:
C₉H₁₁IN₂O |
MDL Number:
MFCD04039866 CAS Number: 470463-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...