(S)-Hexahydropyridazine-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A372180-1G
A372180-250MG
A372180-100MG
77167-968EA
854.99
USD
77167-968
77167-970
77892-768
(S)-Hexahydropyridazine-3-carboxylic acid 95%
(S)-Hexahydropyridazine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₁₀N₂O₂ |
MDL Number:
MFCD19229762 CAS Number: 64044-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...