Benzyl 4-aminopiperidine-1-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A369628-1G A369628-250MG
77167-944EA 19.57 USD
77167-944 77167-946
Benzyl 4-aminopiperidine-1-carboxylate hydrochloride 97%
Benzyl 4-aminopiperidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₉ClN₂O₂
MW: 270.76 g/mol
MDL Number: MFCD09749838
CAS Number: 1159826-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR