3,3-Difluorocyclopentaecarboxylic acid 96%
Supplier: Ambeed
|
Warning
|
A369022-10G
A369022-250MG
A369022-100MG
77167-936EA
1287.5
USD
77167-936
77167-938
77684-071
3,3-Difluorocyclopentaecarboxylic acid 96%
3,3-Difluorocyclopentaecarboxylic acid
Precaución: For research use only.
Formula:
C₆H₈F₂O₂ |
MDL Number:
MFCD15526661 CAS Number: 1260897-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...