Benzamidoxime 98%

Supplier: Ambeed
Danger

A368784-100G A368784-1G A368784-250MG
77195-474EA 140 USD
77195-474 77195-476 77195-478
Benzamidoxime 98%
Benzamidoxime

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MDL Number: MFCD00031485
CAS Number: 613-92-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR