4-(Ethylsulfonyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-(Ethylsulphonyl)phenyl)boronic acid, 4-(Ethylsulphonyl)benzeneboronic acid

A368695-1G A368695-250MG A368695-5G
77195-468EA 67.98 USD
77195-468 77195-470 77195-472
4-(Ethylsulfonyl)phenylboronic acid 97%
4-(Ethylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄S
MW: 214.05 g/mol
MDL Number: MFCD02093076
CAS Number: 352530-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR