1,4-Dibromo-2,5-diiodobenzene 95%

Supplier: Ambeed
Warning

A368346-100G A368346-1G A368346-25G A368346-5G
77195-460EA 255 USD
77195-460 77195-462 77195-464 77195-466
1,4-Dibromo-2,5-diiodobenzene 95%
1,4-Dibromo-2,5-diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂I₂
MW: 487.6970 g/mol
MDL Number: MFCD00082898
CAS Number: 63262-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR