Ethyl viologen dibromide 97%

Supplier: Ambeed
Warning

Synonyms: 1,1'-Diethyl-[4,4'-bipyridine]-1,1'-diium bromide, 1,1'-Diethyl-4,4'-bipyridinium dibromide

A368269-1G A368269-5G
77195-456EA 28 USD
77195-456 77195-458
Ethyl viologen dibromide 97%
Ethyl viologen dibromide

Precaución: for Research Use Only

Formula: C₁₄H₁₈Br₂N₂
MDL Number: MFCD00134590
CAS Number: 53721-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR