3,5-Dimethoxyphenol 96%

Supplier: Ambeed
Warning

A368053-100G A368053-10G A368053-1G A368053-25G A368053-5G
77195-446EA 231 USD
77195-446 77195-448 77195-450 77195-452 77195-454
3,5-Dimethoxyphenol 96%
3,5-Dimethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MDL Number: MFCD00008388
CAS Number: 500-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR