1-Benzyl-2-(tert-butyl)hydrazine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl 2-(tert-butyl) hydrazine-1,2-dicarboxylate, Hydrazine, N-BOC, N'-CBZ protected

A367907-1G A367907-250MG A367907-5G A367907-10G
77195-442EA 22.66 USD
77195-442 77195-444 77616-608 77746-738
1-Benzyl-2-(tert-butyl)hydrazine-1,2-dicarboxylate 97%
1-Benzyl-2-(tert-butyl)hydrazine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₄
MW: 266.2900 g/mol
MDL Number: MFCD09752862
CAS Number: 57699-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR