2-(Ethylthio)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Ethylthio)benzeneboronic acid

A367663-1G A367663-250MG
77195-438EA 88.58 USD
77195-438 77195-440
2-(Ethylthio)phenylboronic acid 97%
2-(Ethylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₂S
MDL Number: MFCD03095256
CAS Number: 362045-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR