N1,N2-Didodecyl-N1,N1,N2,N2-tetramethylethane-1,2-diaminium bromide 97%

Supplier: Ambeed
Warning

A367005-100MG A367005-1G A367005-250MG A367005-5G
77167-914EA 11 USD
77167-914 77167-916 77167-918 77167-920
N1,N2-Didodecyl-N1,N1,N2,N2-tetramethylethane-1,2-diaminium bromide 97%
N1,N2-Didodecyl-N1,N1,N2,N2-tetramethylethane-1,2-diaminium bromide

Precaución: for Research Use Only

Formula: C₃₀H₆₆Br₂N₂
MDL Number: MFCD01861906
CAS Number: 18464-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR