(3-Methyl-1H-pyrazol-5-yl)methanol 97%

Supplier: Ambeed
Warning

A366390-1G A366390-250MG A366390-5G
77195-410EA 34 USD
77195-410 77195-412 77746-733
(3-Methyl-1H-pyrazol-5-yl)methanol 97%
(3-Methyl-1H-pyrazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₈N₂O
MDL Number: MFCD00159583
CAS Number: 17607-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR