4'-(1-Pyrazolyl)acetophenone 97%

Supplier: Ambeed
Warning

A366236-10G A366236-1G A366236-5G
77195-406EA 47 USD
77195-406 77195-408 77746-731
4'-(1-Pyrazolyl)acetophenone 97%
4'-(1-Pyrazolyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MDL Number: MFCD08572143
CAS Number: 25699-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR