2,3-Difluoro-4-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A365450-10G A365450-1G A365450-250MG A365450-5G
77167-906EA 128.75 USD
77167-906 77167-908 77167-910 77167-912
2,3-Difluoro-4-methylbenzonitrile 98%
2,3-Difluoro-4-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MW: 153.13 g/mol
MDL Number: MFCD06660243
CAS Number: 508203-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR