Methyl 2-iodo-3-methylbenzoate 98%
Supplier: Ambeed
|
Warning
|
A365023-100MG
A365023-1G
A365023-250MG
A365023-5G
77167-898EA
19.57
USD
77167-898
77167-900
77167-902
77167-904
Methyl 2-iodo-3-methylbenzoate 98%
Methyl 2-iodo-3-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₉IO₂ MW: 276.071 g/mol |
MDL Number:
MFCD12923249 CAS Number: 103441-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...