1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) 98%

Supplier: Ambeed
Warning

Synonyms: 1,4,7,10,13-Pentaoxa-16-azacyclooctadecane, 1-Aza-18-crown 6-Ether, 1-Aza-18-crown-6

A364824-1G A364824-250MG A364824-5G
77195-398EA 148.32 USD
77195-398 77195-400 77195-402
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) 98%
1-Aza-18-crown-6 (1,4,7,10,13-Pentaoxa-16-azacyclooctadecane)

Precaución: for Research Use Only

Formula: C₁₂H₂₅NO₅
MW: 263.33 g/mol
MDL Number: MFCD00075466
CAS Number: 33941-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR