Methyl 5-hydroxy-6-methoxynicotinate 97%

Supplier: Ambeed
Warning

A364341-100MG A364341-250MG
77167-890EA 57 USD
77167-890 77167-892
Methyl 5-hydroxy-6-methoxynicotinate 97%
Methyl 5-hydroxy-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MDL Number: MFCD16621825
CAS Number: 166742-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR