1-(Trifluoromethyl)vinylboronic acid hexylene glycol ester 98% stabilized

Supplier: Ambeed
Warning

Synonyms: 4,4,6-Trimethyl-2-(3,3,3-trifluoroprop-1-en-2-yl)-1,3,2-dioxaborinane

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A364230-1G A364230-250MG A364230-5G A364230-25G
77167-884EA 27.81 USD
77167-884 77167-886 77167-888 77551-056
1-(Trifluoromethyl)vinylboronic acid hexylene glycol ester 98% stabilized
1-(Trifluoromethyl)vinylboronic acid hexylene glycol ester

Precaución: for Research Use Only

Formula: C₉H₁₄BF₃O₂
MW: 222.012 g/mol
MDL Number: MFCD08669623
CAS Number: 1011460-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR