Methyl-2-acetamidopropionate 95%

Supplier: Ambeed
Warning

Synonyms: N-Acetyl-DL-alanine Methyl Ester, Methyl 2-acetamidopropanoate, 2-Acetamidopropionic Acid Methyl Ester

A363758-1G A363758-5G
77167-880EA 9 USD
77167-880 77167-882
Methyl-2-acetamidopropionate 95%
Methyl-2-acetamidopropionate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃
MW: 145.16 g/mol
MDL Number: MFCD01631289
CAS Number: 26629-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR