(S)-2-Hydroxyethyl 2-amino-3-methylbutanoate 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

A363393-100MG A363393-1G A363393-250MG
77167-874EA 97.85 USD
77167-874 77167-876 77167-878
(S)-2-Hydroxyethyl 2-amino-3-methylbutanoate 4-methylbenzenesulfonate 95%
(S)-2-Hydroxyethyl 2-amino-3-methylbutanoate 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₆S
MW: 333.40 g/mol
MDL Number: MFCD22666220
CAS Number: 86150-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR