2,2,2-Trichloro-1-{1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2,2-Trichloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)ethanone
A363347-100MG
A363347-250MG
A363347-50MG
77167-868EA
69.01
USD
77167-868
77167-870
77167-872
2,2,2-Trichloro-1-{1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one 95%
2,2,2-Trichloro-1-{1H-pyrrolo[2,3-b]pyridin-3-yl}ethan-1-one
Precaución: for Research Use Only
Formula:
C₉H₅Cl₃N₂O MW: 263.51 g/mol |
MDL Number:
MFCD18642801 CAS Number: 163220-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...