(3,5-Difluoro-4-propoxyphenyl)boronic acid 97%
Supplier: Ambeed
|
Warning
|
A363125-100MG
A363125-1G
A363125-250MG
A363125-5G
77195-378EA
35.02
USD
77195-378
77195-380
77195-382
77195-384
(3,5-Difluoro-4-propoxyphenyl)boronic acid 97%
(3,5-Difluoro-4-propoxyphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₁BF₂O₃ MW: 215.99 g/mol |
MDL Number:
MFCD20231441 CAS Number: 2096331-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...