(R)-2-(((Benzyloxy)carbonyl)(methyl)amino)-4-methylpentanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Cbz-D-N-Me-Leu-OH

A362881-10G A362881-25G A362881-5G
77195-372EA 97.85 USD
77195-372 77195-374 77195-376
(R)-2-(((Benzyloxy)carbonyl)(methyl)amino)-4-methylpentanoic acid 98%
(R)-2-(((Benzyloxy)carbonyl)(methyl)amino)-4-methylpentanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.33 g/mol
MDL Number: MFCD04973893
CAS Number: 65635-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR