(tert-Butyldimethylsiloxy)acetaldehyde 90% (by GC)

Supplier: Ambeed
Danger

A362761-5G A362761-10G A362761-1G A362761-250MG A362761-25G
77195-364EA 90 USD
77195-364 77551-046 77551-048 77601-604 77601-606
(tert-Butyldimethylsiloxy)acetaldehyde 90% (by GC)
(tert-Butyldimethylsiloxy)acetaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₈O₂Si
MDL Number: MFCD01321229
CAS Number: 102191-92-4
UN: 1989
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR