2-(4-Acetamidophenoxy)-N,N,N-trimethylethanaminium bromide 97%

Supplier: Ambeed
Warning

A361830-1G A361830-250MG
77167-856EA 33.99 USD
77167-856 77167-858 77167-860
2-(4-Acetamidophenoxy)-N,N,N-trimethylethanaminium bromide 97%
2-(4-Acetamidophenoxy)-N,N,N-trimethylethanaminium bromide

Precaución: for Research Use Only

Formula: C₁₃H₂₁BrN₂O₂
MDL Number: MFCD01674522
CAS Number: 855945-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR