Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Isopropyl 2-(((S)-(perfluorophenoxy)(phenoxy)phosphoryl)amino)propanoate

A361655-100G A361655-10G A361655-25G A361655-5G
77167-850EA 91.67 USD
77167-850 77167-852 77167-854 77551-038
Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate 98%
Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate

Precaución: for Research Use Only

Formula: C₁₈H₁₇F₅NO₅
MW: 453.298 g/mol
MDL Number: MFCD22665799
CAS Number: 1334513-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR