tert-Butyl bis(oxazol-4-ylmethyl)carbamate 97%

Supplier: Ambeed
Warning

A361102-1G A361102-250MG
77167-844EA 24.72 USD
77167-844 77167-846 77167-848
tert-Butyl bis(oxazol-4-ylmethyl)carbamate 97%
tert-Butyl bis(oxazol-4-ylmethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₃O₄
MDL Number: MFCD28369487
CAS Number: 1646152-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR