3-(3-Pyridyl)-L-alanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-Amino-3-(pyridin-3-yl)propanoic acid, H-β-(3-Pyridyl)-Ala-OH, H-3-Pal-OH
A360671-1G
A360671-250MG
A360671-5G
77195-350EA
47.38
USD
77195-350
77195-352
77195-354
3-(3-Pyridyl)-L-alanine 98%
3-(3-Pyridyl)-L-alanine
Precaución: for Research Use Only
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol |
MDL Number:
MFCD00079676 CAS Number: 64090-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...