(R)-2-((S)-6,7-dimethoxy-1-(4-methylphenethyl)-3,4-dihydroisoquinolin-2(1H)-yl)-N-methyl-2-phenylacetamide hydrochloride 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Almorexant HCl
A360660-10MG
A360660-1MG
A360660-25MG
A360660-2MG
A360660-5MG
77167-822EA
48.41
USD
77167-822
77167-824
77167-826
77167-828
77167-830
(R)-2-((S)-6,7-dimethoxy-1-(4-methylphenethyl)-3,4-dihydroisoquinolin-2(1H)-yl)-N-methyl-2-phenylacetamide hydrochloride 98+%
(R)-2-((S)-6,7-dimethoxy-1-(4-methylphenethyl)-3,4-dihydroisoquinolin-2(1H)-yl)-N-methyl-2-phenylacetamide hydrochloride
Precaución: for Research Use Only
Formula:
C₂₉H₃₅ClN₂O₃ MW: 495.05 g/mol |
MDL Number:
MFCD14636620 CAS Number: 913358-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...