4'-Formylbiphenyl-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A360323-100MG A360323-1G A360323-250MG A360323-5G
77167-812EA 35 USD
77167-812 77167-814 77167-816 77167-818
4'-Formylbiphenyl-4-carboxylic acid 98%
4'-Formylbiphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₃
MDL Number: MFCD03424613
CAS Number: 70916-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR