(E)-3-(3,4,5-Trimethoxyphenyl)acrylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3,4,5-Trimethoxycinnamic acid

A359956-100G A359956-10G A359956-5G A359956-500G A359956-25G
77195-336EA 54.59 USD
77195-336 77195-338 77195-340 77746-712 77746-711
(E)-3-(3,4,5-Trimethoxyphenyl)acrylic acid 98%
(E)-3-(3,4,5-Trimethoxyphenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₅
MW: 238.23 g/mol
MDL Number: MFCD00004388
CAS Number: 20329-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR