1,3-Diaminopropane dihydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: Propane-1,3-diamine dihydrochloride, 1,3-Propanediamine dihydrochloride, Trimethylenediamine dihydrochloride

A359674-25G A359674-5G
77195-332EA 47 USD
77195-332 77195-334
1,3-Diaminopropane dihydrochloride 98%
1,3-Diaminopropane dihydrochloride

Precaución: for Research Use Only

Formula: C₃H₁₂Cl₂N₂
MDL Number: MFCD00012525
CAS Number: 10517-44-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR