(3-Ethoxy-4-fluoro-5-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A359652-1G A359652-250MG A359652-5G
77195-326EA 133.9 USD
77195-326 77195-328 77195-330
(3-Ethoxy-4-fluoro-5-(trifluoromethyl)phenyl)boronic acid 97%
(3-Ethoxy-4-fluoro-5-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BF₄O₃
MW: 251.97 g/mol
MDL Number: MFCD20529447
CAS Number: 2096341-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR