2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-silolo[3,2-b:4,5-b']dithiophene 97%

Supplier: Ambeed
Warning

A359517-100MG A359517-1G A359517-250MG
77167-804EA 37 USD
77167-804 77167-806 77167-808
2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-silolo[3,2-b:4,5-b']dithiophene 97%
2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-silolo[3,2-b:4,5-b']dithiophene

Precaución: for Research Use Only

Formula: C₂₄H₃₆Br₂S₂Si
MDL Number: MFCD16621138
CAS Number: 1089687-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR