2-Isopropoxy-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Isopropoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Isopropoxypyridine-3-boronic acid pinacol ester, 2-Isopropoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A359226-1G A359226-250MG A359226-5G
77167-794EA 96 USD
77167-794 77167-796 77167-798
2-Isopropoxy-3-pyridineboronic acid pinacol ester 98%
2-Isopropoxy-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₃
MDL Number: MFCD07366722
CAS Number: 848243-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR