1,3-Diacetoxyacetone 98%

Supplier: Ambeed
Warning

A358294-5G A358294-100G A358294-1G A358294-250MG
77167-788EA 38 USD
77167-788 77892-731 77892-732 77892-733
1,3-Diacetoxyacetone 98%
1,3-Diacetoxyacetone

Precaución: for Research Use Only

Formula: C₇H₁₀O₅
MDL Number: MFCD01310655
CAS Number: 6946-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR